Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rucaparib camsylate
GREENER_CHOICESDP

Catalog No. 623050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-305-0 50 mg
62-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-305-0 Supplier Tocris Bioscience™ Supplier No. 6230/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

PARP inhibitor

Rucaparib camsylate is a PARP inhibitor. Chemosensitizes colorectal cancer cells to Temozolomide (Cat. No. 2706) and Topotecan (Cat. No. 4562) in vitro and in vivo.

Specifications

Chemical Name or Material 8-Fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6 H-pyrrolo[4,3,2,ef][2 ]benzazepin-6-one (1 S,4 R)-7,7-dimethyl-2-oxobicyclo[2.2.1 ]heptane-1-methanesulfonic acid
CAS 1859053-21-6
Assay Percent Range >98%
Quantity 50 mg
Target Poly(ADP-ribose) Polymerase Inhibitors
Molecular Formula C19H18FN3O.C10H16O4S
Formula Weight Observed MW: 555.66
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.