Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RuCl[(R,R)-Tsdpen](mesitylene) 90.0+%, TCI America™
SDP

Catalog No. R01271G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 174813-82-2
Molecular Formula C30H34ClN2O2RuS
Molecular Weight (g/mol) 623.194
MDL Number MFCD12545951
InChI Key BABSEZBJIPZYQB-AGEKDOICSA-M
Synonym [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(mesitylene)ruthenium(II), Chloro(mesitylene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 131675324
IUPAC Name N-[(1R,2R)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide;ruthenium(1+);1,3,5-trimethylbenzene;chloride
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(C2=CC=CC=C2)C(C3=CC=CC=C3)N.CC1=CC(=CC(=C1)C)C.[Cl-].[Ru+]

Specifications

Quantity 1 g
Formula Weight 622.19
Percent Purity ≥90% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material RuCl[(R,R)-Tsdpen](mesitylene)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.