Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rufinamide 98.0+%, TCI America™
SDP

Catalog No. R0143100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mg
100 mg
1 g

Chemical Identifiers

CAS 106308-44-5
Molecular Formula C10H8F2N4O
Molecular Weight (g/mol) 238.198
MDL Number MFCD00865314
InChI Key POGQSBRIGCQNEG-UHFFFAOYSA-N
Synonym rufinamide, inovelon, banzel, 1-2,6-difluorobenzyl-1h-1,2,3-triazole-4-carboxamide, xilep, 1-2,6-difluorophenyl methyl-1h-1,2,3-triazole-4-carboxamide, unii-wfw942pr79, 1-2,6-difluorophenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 1-2,6-difluorophenyl methyl, dsstox_cid_26506
PubChem CID 129228
IUPAC Name 1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
SMILES C1=CC(=C(C(=C1)F)CN2C=C(N=N2)C(=O)N)F

Specifications

Melting Point 241°C
Color White
Quantity 100 mg
Formula Weight 238.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rufinamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.