Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rufinamide
GREENER_CHOICESDP

Catalog No. 382810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-281-0 10 mg
38-285-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-281-0 Supplier Tocris Bioscience™ Supplier No. 3828/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Prolongs inactivation of sodium channels; anticonvulsant

Board spectrum anticonvulsant. Prolongs the inactivation of sodium channels and limits the frequency of action potential firing in cultured and acutely isolated neurons. Displays anticonvulsive activity in a range of animal seizure models.

Chemical Identifiers

CAS 106308-44-5
Molecular Formula C10H8F2N4O

Specifications

Quantity 10 mg
Formula Weight 238.19
Purity >99%
Molecular Weight (g/mol) 238.198
Chemical Name or Material Rufinamide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.