Learn More
Invitrogen™ RUFY1 Polyclonal Antibody

Description
Immunogen sequence: NLQMFHKAQN AESSLQQKNE AITSFEGKTN QVMSSMKQME ERLQHSERAR QGAEERSHKL QQELGGRIGA LQLQLSQLHE QCSSLEKEL Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 84%.
Specifications
Specifications
| Antigen | RUFY1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RUFY1 |
| Gene Accession No. | Q96T51 |
| Gene Alias | 3000002E04Rik; FYVE-finger containing protein; FYVE-finger protein EIP1; La-binding protein 1; Rab4a interacting protein; rab4-interacting protein; Rabip4; RUFY1; RUN and FYVE domain containing 1; RUN and FYVE domain-containing protein 1; ZFYVE12; zinc finger FYVE domain-containing protein 12 |
| Gene Symbols | RUFY1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.