Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUFY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117107 Supplier Invitrogen™ Supplier No. PA5117107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse kidney, Mouse heart, Rat lung Immunogen sequence: EKDTHEKQDT LVALRQQLEE VKAINLQMFH KAQNAESSLQ QKNEAITSFE GKTNQVMSSM KQMEERLQHS ERARQGAEER SHKLQQELGG RIGALQLQLS QLHEQCSSLE KELKSEKEQR QALQRELQHE KDTSSLLRME LQQVEGLKKE LRELQDEKAE LQKICEEQEQ ALQEMGLHLS QSKLKMEDIK EVNQALKGHA WLKDDEATHC RQCEKEFSIS RRKHHCRNCG HIFCNTCSSN ELALPSYPKP VRVCDSCHTL LLQRCSSTAS.

This gene encodes a protein that contains a RUN domain and a FYVE-type zinc finger domain. The encoded protein binds to phosphatidylinositol-3-phosphate (PI3P) and plays a role in early endosomal trafficking, tethering and fusion through interactions with small GTPases including Rab4, Rab5 and Rab14. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUFY1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RUFY1
Gene Accession No. Q8BIJ7
Gene Alias 3000002E04Rik; FYVE-finger containing protein; FYVE-finger protein EIP1; La-binding protein 1; Rab4a interacting protein; rab4-interacting protein; Rabip4; RUFY1; RUN and FYVE domain containing 1; RUN and FYVE domain-containing protein 1; ZFYVE12; zinc finger FYVE domain-containing protein 12
Gene Symbols RUFY1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 439-708 of human RUFY1 (NP_0794343).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216724, 360521
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.