Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUFY1 Polyclonal Antibody
RUFY1 Polyclonal Antibody
Supplier: Thermo Scientific PA531399
Description
RUFY1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
RUFY1 | |
Polyclonal | |
Unconjugated | |
RUFY1 | |
3000002E04Rik; FYVE-finger containing protein; FYVE-finger protein EIP1; La-binding protein 1; Rab4a interacting protein; rab4-interacting protein; Rabip4; RUFY1; RUN and FYVE domain containing 1; RUN and FYVE domain-containing protein 1; ZFYVE12; zinc finger FYVE domain-containing protein 12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80230 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96T51 | |
RUFY1 | |
Recombinant fragment corresponding to a region within amino acids 210 and 529 of Human RUFY1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction