Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RUFY1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531400
Description
Recommended positive controls: IMR32, U87-MG, MCF-7, mouse adipose. Predicted reactivity: Mouse (91%), Rat (91%), Chicken (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that contains a RUN domain and a FYVE-type zinc finger domain. The encoded protein binds to phosphatidylinositol-3-phosphate (PI3P) and plays a role in early endosomal trafficking, tethering and fusion through interactions with small GTPases including Rab4, Rab5 and Rab14. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
RUFY1 | |
Polyclonal | |
Unconjugated | |
RUFY1 | |
3000002E04Rik; FYVE-finger containing protein; FYVE-finger protein EIP1; La-binding protein 1; Rab4a interacting protein; rab4-interacting protein; Rabip4; RUFY1; RUN and FYVE domain containing 1; RUN and FYVE domain-containing protein 1; ZFYVE12; zinc finger FYVE domain-containing protein 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216724, 80230 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8BIJ7, Q96T51 | |
RUFY1 | |
Recombinant protein encompassing a sequence within the center region of human RUFY1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction