Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUFY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576318 Supplier Invitrogen™ Supplier No. PA576318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUFY2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RUFY2
Gene Accession No. Q8R4C2, Q8WXA3
Gene Alias 2610111M19Rik; AI844453; antigen MU-RMS-40.17; Denn; Kiaa1537; Leucine zipper FYVE-finger protein; LZ-FYVE; Rab4-interacting protein related; RABIP4R; RUFY2; RUN and FYVE domain containing 2; RUN and FYVE domain-containing 2; RUN and FYVE domain-containing protein 2; Run- and FYVE-domain containing protein; ZFYVE13
Gene Symbols RUFY2
Host Species Rabbit
Immunogen Recombinant protein of human RUFY2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55680, 690777, 70432
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.