Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUFY4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567535 Supplier Invitrogen™ Supplier No. PA567535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specificity: The antibody detects endogenous levels of total RUFY4 protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RUFY4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RUFY4
Gene Accession No. Q6ZNE9
Gene Alias RUFY4; RUN and FYVE domain containing 4; RUN and FYVE domain-containing protein 4; ZFYVE31
Gene Symbols RUFY4
Host Species Rabbit
Immunogen Fusion protein of human RUFY4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285180
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.