Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571941
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571941 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571941 Supplier Invitrogen™ Supplier No. PA571941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RUNDC1 Polyclonal Antibody for Western Blot

RUNDC1 may play a role as p53/TP53 inhibitor and thus may have oncogenic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNDC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RUNDC1
Gene Accession No. Q0VDN7, Q96C34
Gene Alias AW125546; D930024B17; RUN domain containing 1; RUN domain-containing protein 1; RUNDC1
Gene Symbols RUNDC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 509-537 amino acids from the C-terminal region of human RUNDC1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146923, 217201
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.