Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNDC3A Monoclonal Antibody (1A8)
GREENER_CHOICE

Catalog No. PIMA521185
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521185 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521185 Supplier Invitrogen™ Supplier No. MA521185
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEASFVQTTM ALGLSSKKAS SRNVAVERKN LITVCRFSVK TLLEKYTAEP IDDSSEEFVN FAAILEQILS HRFKACAPAG PVSWFSSDGQ RGFWDYIRLA CSKVPNNCVS SIENMENIST ARAKGRAWIR VALMEKRMSE YITTALRDTR TTRRFYDSGA IMLRDEATIL TGMLIGLSAI DFSFCLKGEV LDGKTPVVID YTPYLKFTQS YDYLTDEEER YSAESSTSED NSPEHPYLPL VTDEDSWYSK WHKMEQKFRI VYAQKGYLEE LVRLRESQLK DLEAENRRLQ LQLEEAAAQN QREKRELEGV ILELQEQLTG LIPSDHAPLA QGSKELTTPL VNQWPSLGTL NGAEGASNSK LYRRHSFMST EPLSAEASLS SDSQRLGEGT RDEEPWGPIG SSEPN.

May act as an effector of RAP2A in neuronal cells. Classified as a regulatory protein which binds to guanylate cyclase GC-A and potentiates its activation by atrial natriuretic factor and ATP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNDC3A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RUNDC3A
Gene Accession No. Q59EK9
Gene Alias GCRP; guanylate-cyclase regulatory protein; Rap2 interacting protein; Rap2 interacting protein 8; rap2-interacting protein 8; Rap2ip; Rpip8; RPIP-8; RUN domain containing 3A; RUN domain-containing protein 3A; Rundc3a
Gene Symbols RUNDC3A
Host Species Mouse
Immunogen RUNDC3A (AAH06194, 1 a.a. ~ 405 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10900, 303569
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.