Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNDC3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584848 Supplier Invitrogen™ Supplier No. PA584848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDEEERHSAE SSTSEDNSPE HPYLPLVTDE DSWYSKWHKM EQKFRIVYAQ K Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

May act as an effector of RAP2A in neuronal cells. Classified as a regulatory protein which binds to guanylate cyclase GC-A and potentiates its activation by atrial natriuretic factor and ATP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNDC3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RUNDC3A
Gene Accession No. Q59EK9
Gene Alias GCRP; guanylate-cyclase regulatory protein; Rap2 interacting protein; Rap2 interacting protein 8; rap2-interacting protein 8; Rap2ip; Rpip8; RPIP-8; RUN domain containing 3A; RUN domain-containing protein 3A; Rundc3a
Gene Symbols RUNDC3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RUNDC3A. Recombinant protein control fragment (Product #RP-108572).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10900
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.