Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNDC3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598472
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598472 Supplier Invitrogen™ Supplier No. PA598472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RUNDC3A Polyclonal Antibody for Western Blot, IHC (P), ELISA

May act as an effector of RAP2A in neuronal cells. Classified as a regulatory protein which binds to guanylate cyclase GC-A and potentiates its activation by atrial natriuretic factor and ATP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNDC3A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RUNDC3A
Gene Accession No. Q59EK9
Gene Alias GCRP; guanylate-cyclase regulatory protein; Rap2 interacting protein; Rap2 interacting protein 8; rap2-interacting protein 8; Rap2ip; Rpip8; RPIP-8; RUN domain containing 3A; RUN domain-containing protein 3A; Rundc3a
Gene Symbols RUNDC3A
Host Species Rabbit
Immunogen Recombinant Human RUN domain-containing protein 3A protein (1-405AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10900
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.