Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUNX1/CBFA2 Antibody (6C12A5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$541.95
Specifications
| Antigen | RUNX1/CBFA2 |
|---|---|
| Clone | 6C12A5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1-2000, Flow Cytometry 1:200-1:400, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61861
![]() |
Novus Biologicals
NBP261861 |
0.1 mL |
Each of 1 for $541.95
|
|
|||||
Description
RUNX1/CBFA2 Monoclonal specifically detects RUNX1/CBFA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| RUNX1/CBFA2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Acute myeloid leukemia 1 protein, AML1acute myeloid leukemia 1, AML1-EVI-1, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, Core-binding factor subunit alpha-2, EVI-1, PEBP2A2, runt domain, alpha subunit 2, runt-related transcription factor 1, SL3/AKV core-binding factor alpha B subunit | |
| RUNX1 | |
| IgG1 | |
| Protein G purified |
| 6C12A5 | |
| Western Blot 1:500-1-2000, Flow Cytometry 1:200-1:400, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Cancer, Phospho Specific, Tumor Suppressors | |
| 861 | |
| Purified recombinant fragment of human RUNX1 (AA: 237-337) expressed in E. Coli. | |
| Primary | |
| 48.7 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title