Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1/CBFA2 Antibody (6C12A5) - BSA Free, R&D Systems™
SDP

Catalog No. NBP261861 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61861 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61861 Supplier R&D Systems Supplier No. NBP261861

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

RUNX1/CBFA2 Monoclonal specifically detects RUNX1/CBFA2 in Human samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen RUNX1/CBFA2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6C12A5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1-2000, Flow Cytometry 1:200-1:400, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias Acute myeloid leukemia 1 protein, AML1acute myeloid leukemia 1, AML1-EVI-1, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, Core-binding factor subunit alpha-2, EVI-1, PEBP2A2, runt domain, alpha subunit 2, runt-related transcription factor 1, SL3/AKV core-binding factor alpha B subunit
Gene Symbols RUNX1
Host Species Mouse
Immunogen Purified recombinant fragment of human RUNX1 (AA: 237-337) expressed in E. Coli.
Molecular Weight of Antigen 48.7 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 861
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.