Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1/CBFA2 Antibody, Novus Biologicals™
SDP

Catalog No. p-5089923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002044SS 0.025 mg
NB1002044 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002044SS Supplier Novus Biologicals Supplier No. NB1002044SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RUNX1/CBFA2 Polyclonal specifically detects RUNX1/CBFA2 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen RUNX1/CBFA2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q01196
Gene Alias Acute myeloid leukemia 1 protein, AML1acute myeloid leukemia 1, AML1-EVI-1, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, Core-binding factor subunit alpha-2, EVI-1, PEBP2A2, runt domain, alpha subunit 2, runt-related transcription factor 1, SL3/AKV core-binding factor alpha B subunit
Gene Symbols RUNX1
Host Species Rabbit
Immunogen Rabbit anti-AML1 polyclonal antibody was raised against a peptide corresponding to a sequence that is homologous in AML1 and AML1/ETO (amino acids 231 to 245 of human origin).
Molecular Weight of Antigen 49 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 861
Test Specificity Human and Mouse. Not yet tested in other species.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.