Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1/CBFA2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP255213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP255213 100 μL
NB395117 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP255213 Supplier Novus Biologicals Supplier No. NBP255213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RUNX1/CBFA2 Polyclonal specifically detects RUNX1/CBFA2 in Human samples. It is validated for ChIP Assay-exo-Seq, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RUNX1/CBFA2
Applications ChIP Assay-exo-Seq, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias Acute myeloid leukemia 1 protein, AML1acute myeloid leukemia 1, AML1-EVI-1, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, Core-binding factor subunit alpha-2, EVI-1, PEBP2A2, runt domain, alpha subunit 2, runt-related transcription factor 1, SL3/AKV core-binding factor alpha B subunit
Gene Symbols RUNX1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LDDQTKPGSLSFSERLSELEQLRRTAMRVSPHHPAPTPNPRASLNHSTAFN
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 861
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.