Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNX1 Recombinant Rabbit Monoclonal Antibody (HL2211)
GREENER_CHOICE

Catalog No. FEMA550434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550434 Supplier Invitrogen™ Supplier No. MA550434
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

RUNX1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

RUNX1 belongs to the mammalian RUNX family of DNA binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX1 is essential for hematopoietic stem cell generation in the vascular regions of the aorta, vitelline and umbilical arteries, yolk sac and placenta. RUNX1 is expressed in all tissues except brain and heart, with highest levels in thymus, bone marrow and peripheral blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2211
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RUNX1
Gene Accession No. Q01196, Q03347, Q63046
Gene Alias acute myeloid leukemia 1 protein; Acute myeloid leukemia 2 protein; acute myeloid leukemia 3 protein; acute myeloid leukemia gene 2; AML1; AML-1; aml1 oncogene; AML1-EVI-1; AML1-EVI-1 fusion protein; AML2; AML3; AMLCR1; CBF2alpha; CBFA1; CBFA2; CBFA3; CBF-alpha 2; CBF-alpha-1; CBF-alpha-2; CBF-alpha-3; CCD; CCD1; CLCD; core binding factor alpha 2; core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene); Core-binding factor subunit alpha-1; Core-binding factor subunit alpha-2; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 1; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 3; EVI-1; Oncogene AML-1; oncogene AML-2; Oncogene AML-3; OSF2; OSF-2; osteoblast-specific transcription factor 2; OTTHUMP00000108697; OTTHUMP00000108700; OTTHUMP00000108702; PEA2 alpha C; PEA2aA; PEA2-alpha A; PEA2-alpha B; PEA2-alpha C; PEBP2 alpha C; PEBP2A; Pebp2a2; PEBP2A3; PEBP2aA; PEBP2aB; PEBP2aC; PEBP2alpha; PEBP2-alpha A; PEBP2-alpha B; PEBP2-alpha C; Pebpa2b; polyomavirus enhancer-binding protein 2 alpha A subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; polyomavirus enhancer-binding protein 2 alpha C subunit; RUN1; runt domain, alpha subunit 2; runt related transcription factor 1; runt related transcription factor 2; runt related transcription factor 3; runt-related transcription factor 1; Runt-related transcription factor 2; runt-related transcription factor 3; Runx1; RUNX1/ZNF687 fusion; RUNX2; RUNX3; SL3/AKV core-binding factor alpha A subunit; SL3/AKV core-binding factor alpha B subunit; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha A subunit; SL3-3 enhancer factor 1 alpha B subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor; transcription factor AML2
Gene Symbols RUNX1
Host Species Rabbit
Immunogen Recombinant fragmemt of human RUNX1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12394, 50662, 861
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.