Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNX1/RUNX2/RUNX3 Recombinant Rabbit Monoclonal Antibody (6G5U2)
GREENER_CHOICE

Catalog No. PIMA535649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535649 Supplier Invitrogen™ Supplier No. MA535649
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: LPPPYPGSSQ AQGGPFQASS PSYHLYYGAS AGSYQFSMVG GERSPPRILP PCTNASTGSA LLNPSLPNQS DVVEAEGSHS NSPTNMAPSA RLEEAVWRPY.

Core binding factor (CBF) is a heterodimeric transcription factor that binds to the core element of many enhancers and promoters. The protein encoded by this gene represents the alpha subunit of CBF and is thought to be involved in the development of normal hematopoiesis. Chromosomal translocations involving this gene are welldocumented and have been associated with several types of leukemia. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX1/RUNX2/RUNX3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6G5U2
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RUNX3
Gene Accession No. Q01196, Q03347, Q08775, Q13761, Q13950, Q64131
Gene Alias acute myeloid leukemia 1 protein; Acute myeloid leukemia 2 protein; acute myeloid leukemia 3 protein; acute myeloid leukemia gene 2; AKV core binding factor; AML1; AML1-EVI-1; AML1-EVI-1 fusion protein; AML2; Aml3; AMLCR1; Cbf; CBF2alpha; Cbfa1; Cbfa-1; CBFA2; Cbfa3; CBF-alpha-1; CBF-alpha-2; CBF-alpha-3; CCD; CCD1; CLCD; core binding factor alpha 1; core binding factor alpha 2; core binding factor alpha 3; core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene); core-binding factor subunit alpha-1; Core-binding factor subunit alpha-2; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 1; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 3; EVI-1; LS3; oncogene AML-1; Oncogene AML-2; oncogene AML-3; OSF2; OSF-2; Osteoblast-specific transcription factor 2; PEA2 alpha C; PEA2aA; PEA2-alpha A; PEA2-alpha B; PEA2-alpha C; PEBP2 alpha A; PEBP2 alpha C; Pebp2a; Pebp2a1; Pebp2a2; Pebp2a3; PEBP2aA; PEBP2aB; PEBP2aC; PEBP2alpha; PEBP2-alpha A; PEBP2-alpha B; PEBP2-alpha C; Pebpa2a; Pebpa2b; Polyomavirus enhancer-binding protein 2 alpha A subunit; polyomavirus enhancer-binding protein 2 alpha B subunit; Polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 1; runt domain, alpha subunit 2; runt domain, alpha subunit 3; runt related transcription factor 1; runt related transcription factor 2; runt related transcription factor 3; runt-related transcription factor 1; Runt-related transcription factor 2; runt-related transcription factor 3; RUNX 2; Runx1; RUNX1/ZNF687 fusion; Runx2; RUNX-2; RUNX3; SL3/AKV core-binding factor alpha A subunit; SL3/AKV core-binding factor alpha B subunit; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha A subunit; SL3-3 enhancer factor 1 alpha B subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor; transcription factor AML2; transcription factor AML2/CBFA3
Gene Symbols RUNX1, Runx2, RUNX3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 354-453 of human RUNX1/2/3 (NP_0010018901).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12393, 12394, 12399, 860, 861, 864
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.