Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUNX1T1/ETO Antibody (OTI1H1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273956AF594
Description
RUNX1T1/ETO Monoclonal specifically detects RUNX1T1/ETO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RUNX1T1/ETO | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
RUNX1T1 | |
Full length recombinant protein of human RUNX1T1/ETO (NP_783553) produced in HEK293T cells. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1H1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
AML1T1protein CBFA2T1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, Protein ETO, Protein MTG8, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22 | |
Mouse | |
0.1 mL | |
862 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction