Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNX1T1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540817
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540817 Supplier Invitrogen™ Supplier No. PA540817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LHSEHPSKRP CTISPGQRYS PNNGLSYQPN GLPHPTPPPP QHYRLDDMAI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

RUNX1T1 is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. Several transcript variants encoding multiple isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX1T1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RUNX1T1
Gene Accession No. Q06455
Gene Alias acute myelogenous leukemia 1 translocation 1 protein; acute myelogenous leukemia 1 translocation 1, cyclin-D related; AML1-MTG8; AML1T1; Cbfa2t1; CBFA2T1 identified gene homolog; CBFA2T1 isoform r1t1-11a61; CBFA2T1 isoform r1t1-11a62; CBFA2T1 isoform r1t1-11a63; CBFA2T1 isoform r1t1-11a64; CBFA2T1 isoform r1t1-11a65; CBFA2T1 isoform r1t1-7a47; CBFA2T1 isoform r1t1-7a48; CBFA2T1 isoform r1t1-7a49; CBFA2T1 isoform r1t1-7a50; CBFA2T1 isoform r1t1-7a51; CBFA2T1 isoform r1t1-7a52; CBFA2T1 isoform r1t1-7d53; CBFA2T1 isoform r1t1-7d54; CBFA2T1 isoform r1t1-7d55; CBFA2T1 isoform r1t1-7d56; CBFA2T1 isoform r1t1-8a57; CBFA2T1 isoform r1t1-8a58; CBFA2T1 isoform r1t1-8a59; CBFA2T1 isoform r1t1-8a60; Cbfa2t1h; CDR; core-binding factor, runt domain, alpha subunit 2, translocated to, 1; core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclin D-related; cyclin D-related; Cyclin-D-related protein; Eight twenty one protein; ETO; LOW QUALITY PROTEIN: protein CBFA2T1; MGC2796; MTG8; MTG8b; myeloid translocation gene on 8q22; protein CBFA2T1; protein CBFA2T1; LOW QUALITY PROTEIN: protein CBFA2T1; protein CBFA2T1-like; Protein ETO; Protein MTG8; runt related transcription factor 1; translocated to, 1 (cyclin D related); runt-related transcription factor 1; translocated to, 1 (cyclin D-related); RUNX1 partner transcriptional co-repressor 1; RUNX1 translocation partner 1; Runx1t1; si:ch211-232j17.1; translocated to, 1; wu:fi14b07; zgc:154044; zinc finger MYND domain-containing protein 2; ZMYND2
Gene Symbols RUNX1T1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RUNX1T1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 862
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.