Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1T1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595911 Supplier Thermo Scientific Supplier No. PA595911
Add to Cart
Add to Cart

RUNX1T1 Polyclonal Antibody

RUNX1T1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX1T1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RUNX1T1
Gene Accession No. Q06455
Gene Alias acute myelogenous leukemia 1 translocation 1 protein; acute myelogenous leukemia 1 translocation 1, cyclin-D related; AML1-MTG8; AML1T1; Cbfa2t1; CBFA2T1 identified gene homolog; CBFA2T1 isoform r1t1-11a61; CBFA2T1 isoform r1t1-11a62; CBFA2T1 isoform r1t1-11a63; CBFA2T1 isoform r1t1-11a64; CBFA2T1 isoform r1t1-11a65; CBFA2T1 isoform r1t1-7a47; CBFA2T1 isoform r1t1-7a48; CBFA2T1 isoform r1t1-7a49; CBFA2T1 isoform r1t1-7a50; CBFA2T1 isoform r1t1-7a51; CBFA2T1 isoform r1t1-7a52; CBFA2T1 isoform r1t1-7d53; CBFA2T1 isoform r1t1-7d54; CBFA2T1 isoform r1t1-7d55; CBFA2T1 isoform r1t1-7d56; CBFA2T1 isoform r1t1-8a57; CBFA2T1 isoform r1t1-8a58; CBFA2T1 isoform r1t1-8a59; CBFA2T1 isoform r1t1-8a60; Cbfa2t1h; CDR; core-binding factor, runt domain, alpha subunit 2, translocated to, 1; core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclin D-related; cyclin D-related; Cyclin-D-related protein; Eight twenty one protein; ETO; LOW QUALITY PROTEIN: protein CBFA2T1; MGC2796; MTG8; MTG8b; myeloid translocation gene on 8q22; protein CBFA2T1; protein CBFA2T1; LOW QUALITY PROTEIN: protein CBFA2T1; protein CBFA2T1-like; Protein ETO; Protein MTG8; runt related transcription factor 1; translocated to, 1 (cyclin D related); runt-related transcription factor 1; translocated to, 1 (cyclin D-related); RUNX1 partner transcriptional co-repressor 1; RUNX1 translocation partner 1; Runx1t1; si:ch211-232j17.1; translocated to, 1; wu:fi14b07; zgc:154044; zinc finger MYND domain-containing protein 2; ZMYND2
Gene Symbols RUNX1T1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-403 of human RUNX1T1 (NP_7835521).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 862
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.