Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUNX1T1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111303
Description
Immunogen sequence: KGGGSSSSHS RQQSPVNPDP VALDAHREFL HRPASGYVPE EIWKKAE
RUNX1T1 is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. Several transcript variants encoding multiple isoforms have been found for this gene.
Specifications
RUNX1T1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q06455 | |
RUNX1T1 | |
Recombinant Protein corresonding to Human RUNX1T1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
Unconjugated | |
RUNX1T1 | |
acute myelogenous leukemia 1 translocation 1 protein; acute myelogenous leukemia 1 translocation 1, cyclin-D related; AML1-MTG8; AML1T1; Cbfa2t1; CBFA2T1 identified gene homolog; CBFA2T1 isoform r1t1-11a61; CBFA2T1 isoform r1t1-11a62; CBFA2T1 isoform r1t1-11a63; CBFA2T1 isoform r1t1-11a64; CBFA2T1 isoform r1t1-11a65; CBFA2T1 isoform r1t1-7a47; CBFA2T1 isoform r1t1-7a48; CBFA2T1 isoform r1t1-7a49; CBFA2T1 isoform r1t1-7a50; CBFA2T1 isoform r1t1-7a51; CBFA2T1 isoform r1t1-7a52; CBFA2T1 isoform r1t1-7d53; CBFA2T1 isoform r1t1-7d54; CBFA2T1 isoform r1t1-7d55; CBFA2T1 isoform r1t1-7d56; CBFA2T1 isoform r1t1-8a57; CBFA2T1 isoform r1t1-8a58; CBFA2T1 isoform r1t1-8a59; CBFA2T1 isoform r1t1-8a60; Cbfa2t1h; CDR; core-binding factor, runt domain, alpha subunit 2, translocated to, 1; core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclin D-related; cyclin D-related; Cyclin-D-related protein; Eight twenty one protein; ETO; LOW QUALITY PROTEIN: protein CBFA2T1; MGC2796; MTG8; MTG8b; myeloid translocation gene on 8q22; protein CBFA2T1; protein CBFA2T1; LOW QUALITY PROTEIN: protein CBFA2T1; protein CBFA2T1-like; Protein ETO; Protein MTG8; runt related transcription factor 1; translocated to, 1 (cyclin D related); runt-related transcription factor 1; translocated to, 1 (cyclin D-related); RUNX1 partner transcriptional co-repressor 1; RUNX1 translocation partner 1; Runx1t1; si:ch211-232j17.1; translocated to, 1; wu:fi14b07; zgc:154044; zinc finger MYND domain-containing protein 2; ZMYND2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction