Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1T1, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89998420 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-420 Supplier Abnova Corporation Supplier No. PAB31305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen RUNX1T1
Applications ChIP Assay, ChIP sequencing (ChIP-seq), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA (1:100) ChIP (4 ug/ChIP) The optimal working dilution should be determined by the end user.
Formulation In whole antiserum (0.05% sodium azide).
Gene runt-related transcription factor 1; translocated to, 1 (cyclin D-related)
Gene Alias AML1T1/CBFA2T1/CDR/ETO/MGC2796/MTG8/MTG8b/ZMYND2
Gene Symbols RUNX1T1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus and the central region of human RUNX1T1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 862
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.