Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUNX2/CBFA1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177462AF532
Description
RUNX2/CBFA1 Polyclonal specifically detects RUNX2/CBFA1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RUNX2/CBFA1 | |
Polyclonal | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Acute myeloid leukemia 3 protein, AML3CCD, CBFA1MGC120022, Core-binding factor subunit alpha-1, MGC120023, PEA2-alpha A, PEBP2A, PEBP2A1CCD1, PEBP2aA1, PEBP2-alpha A, runt domain, alpha subunit 1, runt-related transcription factor 2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit | |
Rabbit | |
56.6 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
RUNX2 | |
This RUNX2/CBFA1 Antibody was prepard from a synthetic peptide made to an internal region of the human RUNX2 protein (within residues 225-300). [Swiss-Prot Q13950] | |
Affinity Purified | |
Phospho Specific, Transcription Factors and Regulators | |
860.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction