Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUNX3/CBFA3 Antibody (K12P3F11*C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | RUNX3/CBFA3 |
---|---|
Clone | K12P3F11*C9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Description
RUNX3/CBFA3 Monoclonal antibody specifically detects RUNX3/CBFA3 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
RUNX3/CBFA3 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Transcription Factors and Regulators | |
PBS | |
864 | |
IgG1 κ | |
Protein A purified |
K12P3F11*C9 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Acute myeloid leukemia 2 protein, AML2SL3/AKV core-binding factor alpha C subunit, CBFA3MGC16070, CBF-alpha-3, PEA2 alpha C, PEA2-alpha C, PEBP2 alpha C, PEBP2A3FLJ34510, PEBP2-alpha C, runt domain, alpha subunit 3, runt-related transcription factor 3, SL3-3 enhancer factor 1 alpha C subunit, transcription factor AML2 | |
Peptide - QFDRSFPTL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title