Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNX3 Monoclonal Antibody (7E2B8)
GREENER_CHOICE

Catalog No. FERMA548407
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548407 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548407 Supplier Invitrogen™ Supplier No. MA548407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RUNX3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5'-PYGPYGGT-3' found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7E2B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RUNX3
Gene Accession No. Q13761
Gene Alias Acute myeloid leukemia 2 protein; acute myeloid leukemia gene 2; AML2; CBFA3; CBF-alpha 3; CBF-alpha-3; core binding factor alpha 3; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 3; oncogene AML-2; PEA2 alpha C; PEA2-alpha C; PEBP2 alpha C; PEBP2A3; PEBP2aC; PEBP2-alpha C; polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 3; Runt related transcription factor 3; runt-related transcription factor 3; RUNX3; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor AML2; transcription factor AML2/CBFA3
Gene Symbols RUNX3
Host Species Mouse
Immunogen Purified recombinant fragment of human RUNX3 (AA: 294-429) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.