Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RUNX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579944
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, Mouse Liver Tissue.
This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5'-PYGPYGGT-3' found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
RUNX3 | |
Polyclonal | |
Unconjugated | |
RUNX3 | |
Acute myeloid leukemia 2 protein; acute myeloid leukemia gene 2; AML2; CBFA3; CBF-alpha 3; CBF-alpha-3; core binding factor alpha 3; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 3; oncogene AML-2; PEA2 alpha C; PEA2-alpha C; PEBP2 alpha C; PEBP2A3; PEBP2aC; PEBP2-alpha C; polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 3; Runt related transcription factor 3; runt-related transcription factor 3; RUNX3; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor AML2; transcription factor AML2/CBFA3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12399, 156726 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q64131 | |
RUNX3 | |
A synthetic peptide corresponding to a sequence in the middle region of mouse RUNX3 (186-205aa QKIEDQTKAFPDRFGDLRMR). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction