Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208915
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8915 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8915 Supplier Fabgennix Inc Supplier No. RUNX3301AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RUNX3 Polyclonal antibody specifically detects RUNX3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen RUNX3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene RUNX3
Gene Accession No. Q13761, Q64131
Gene Alias Acute myeloid leukemia 2 protein; acute myeloid leukemia gene 2; AML2; CBFA3; CBF-alpha 3; CBF-alpha-3; core binding factor alpha 3; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 3; oncogene AML-2; PEA2 alpha C; PEA2-alpha C; PEBP2 alpha C; PEBP2A3; PEBP2aC; PEBP2-alpha C; polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 3; Runt related transcription factor 3; runt-related transcription factor 3; RUNX3; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor AML2; transcription factor AML2/CBFA3
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-50 on human Runt-related transcription factor 3 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12399, 156726, 864
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.