Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ruplizumab Chimeric Recombinant Rabbit Monoclonal Antibody (hu5c8 (Ruplizumab))
GREENER_CHOICE

Catalog No. PIMA547966
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547966 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547966 Supplier Invitrogen™ Supplier No. MA547966
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: The antibody binds to CD40L with an IC50 of 0.845 μg/mL.

Ruplizumab is a humanized monoclonal antibody used as an immunosuppressive pharmaceutical and is a component of Antova. Ruplizumab is an anti-CD40L monoclonal antibody. The binding of CD40 to its ligand, CD40L is a critical element in T cell activation. In systemic lupus erythematosus, CD40L is over-expressed on T cells, B cells and monocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ruplizumab Chimeric
Applications Flow Cytometry, Inhibition Assays, Western Blot
Classification Recombinant Monoclonal
Clone hu5c8 (Ruplizumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Ntrk1
Gene Accession No. P04629
Gene Alias BG-9588; hu5c8
Host Species Rabbit
Immunogen Human CD40L.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4914
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.