Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUSC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556298 Supplier Invitrogen™ Supplier No. PA556298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAMLHFGGRL AQSLRGTSKE AASDPSDSPN LPTPGSWWEQ LTQASRVYAS GGTEGFPLSR WAPGRHGTAA EEGAQERPLP TDEMAPGRGL WLGRLFGVP Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 71%.

RUSC1, also known as NESCA, shares with the related protein RUSC2 a common domain structure of RUN, leucine zipper and SH3 domain in addition to over 30% amino acid identity. RUSC1 is an adapter protein that can bind to the TrkA receptor and is necessary in the NGF-induced neurite growth of PC12 cells. RUSC1 has also been shown to interact with Ikappa-B kinase- (IKK-) -gamma, the regulatory subunit of the IKK complex that is required for NF-kappa-B activation in many signaling pathways such as TNF-R or the TLR pathways. RUSC1 can also bind to the E3 ubiquitin ligase TRAF6, which then catalyzes RUSC1 polyubiquitination. Since overexpression of RUSC1 strongly inhibits TRAF6-mediated polyubiquitination of IKK-gamma, RUSC1 may be a link in the IKK-gamma-mediated NF-kappa-B activation pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUSC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RUSC1
Gene Accession No. Q9BVN2
Gene Alias 2210403N08Rik; AA408288; LOC100360417; Nesca; new molecule containing SH3 at the carboxy-terminus; RP11-21N7.4; RUN and SH3 domain containing 1; RUN and SH3 domain containing 1-like; RUN and SH3 domain-containing protein 1; RUSC1
Gene Symbols RUSC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RUSC1. Recombinant protein control fragment (Product #RP-94358).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.