Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUSC2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281785G
This item is not returnable.
View return policy
Description
RUSC2 Polyclonal antibody specifically detects RUSC2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RUSC2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| interacting protein of Rab1, Iporin, KIAA0375, RUN and SH3 domain containing 2, RUN and SH3 domain-containing protein 2 | |
| RUSC2 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human RUSC2. The immunogen is located within amino acids 1410 - 1460 of RUSC2. Amino Acid Squence: SRREPEPKESLQEPH | |
| 0.1 mL | |
| Signal Transduction | |
| 9853 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction