Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUSC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588854 Supplier Invitrogen™ Supplier No. PA588854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGDVDRAAQG ERVKGVGASE GGEEEEEEEE TEEVAEAAGG SGRARWARGG QAGWWYQLMQ SSQVYIDGSI EGSRFPRGSS NSSSEKKKGA GGGGPPQAPP PREGVVEGAE ACPASEEALG RERGWPFWMG SPPDSVLAEL RRSREREGPA ASPAENEEGA SEPSPGGIKW GHLFGSRKAQ REARPTNRLP SDWLSLDKSM FQLVAQTVGS RREPEPKESL QEPHSPALPS SPPCEVQALC HHLATGPGQL SFHKGDILRV LGRAGGDWLR CSRGPDSGLV PLAYVTLTPT PSPTPGSSQN; Positive Samples: Rat brain; Cellular Location: Cytoplasm, cytosol.

This gene encodes a RUN and SH3 domain containing protein that interacts with Rab1b and Rab1-binding protein GM130. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUSC2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RUSC2
Gene Alias AI840675; Interacting protein of Rab1; iporin; KIAA0375; mKIAA0375; RUN and SH3 domain containing 2; RUN and SH3 domain-containing protein 2; RUSC2
Gene Symbols RUSC2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1217-1516 of human RUSC2 (NP_0556211).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 684259
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.