Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet

Catalog No. AA4433806
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AA4433806

Supplier: Thermo Scientific Chemicals 04433806

Only null left
Add to Cart

CAS: 7440-18-8 | Ru | 101.07 g/mol

Ruthenium, 5% on activated carbon powde is used in solar cells, which turn light energy into electrical energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Ruthenium, 5% on activated carbon powder, Reduced, nominally 50% water wet
EINECSNumber 231-127-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.