Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder

Catalog No. AC198100050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 7440-18-8 | Ru | 101.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Color Gray
Quantity 5 g
Packaging Glass bottle
Form Powder
Assay Percent Range 99.90%
Merck Index 15, 8433
Formula Weight 101.07
Grade Trace Metal
Chemical Name or Material Ruthenium, -200 mesh
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.