Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous

Catalog No. p-4509792
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle

CAS: 10049-08-8 | Cl3Ru | 207.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-08-8
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.43

Specifications

Melting Point >500.0°C
Color Black
Physical Form Crystals or Powder
Quantity 2 g
Assay Percent Range 48 to 50% (Ru)
Linear Formula RuCl3
Fieser 04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292
Merck Index 15,8436
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water, slightly soluble in alcohol
Formula Weight 207.43
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ruthenium(III) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 233-167-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.