Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(III) iodide, anhydrous, Ru 20.5% min

Catalog No. p-7048868
Click to view available options
Quantity:
1 g
5 g

CAS: 13896-65-6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13896-65-6
MDL Number MFCD00016316
Synonym ruthenium iii iodide, ruthenium iii ; iodide, ruthenium 3+ triiodide, ruthenium iii iodide anhydrous, ruthenium iii iodide, anhydrous 1g, ruthenium iii iodide, anhydrous, ru min

Specifications

Melting Point 590°C (decomposition)
Physical Form Powder
Quantity 1 g
Formula Weight 481.78
Concentration or Composition (by Analyte or Components) Ru 20.5% min.
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) iodide, Anhydrous
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-664-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.