Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(III) nitrosylchloride, Ru 42.1% min

Catalog No. p-7049629
Click to view available options
Quantity:
1 g
5 g

CAS: 18902-42-6 | Cl3NORu | 237.43 g/mol

Ruthenium nitrosyl chloride is an important ruthenium compounds, which is used in synthesis of more complex ruthenium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18902-42-6
Molecular Formula Cl3NORu
Molecular Weight (g/mol) 237.43
MDL Number MFCD00049765
InChI Key AUWSZCCJRSCQEP-UHFFFAOYSA-K

Specifications

Physical Form Crystalline
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 237.44
Concentration or Composition (by Analyte or Components) Ru 42.1% min.
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) nitrosylchloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-651-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.