Ruthenium(III) nitrosylnitrate, Ru 31.3% min., Alfa Aesar™

Manufacturer: Alfa Aesar 1217514

Quantity 25g

 View more versions of this product

Catalog No. AA1217514

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 34513-98-9
    Physical Form Crystalline
    Assay Percent Range Ru 31.3% min.
    Molecular Formula N4O10Ru
    Formula Weight 317.09
    MDL Number MFCD00016313
    Chemical Name or Material Ruthenium(III) nitrosylnitrate

    Specifications

    Formula Weight 317.09
    Quantity 25g
    Color Red-brown
    DOT Information DOT Class: 5.1, Packaging Group: II
    EINECS Number 252-068-8
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range Ru 31.3% min.
    Molecular Formula N4O10Ru
    Appearance Red-brown crystals
    MDL Number MFCD00016313
    CAS 34513-98-9
    Health Hazard 1 H272-H314-H318-H290: May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals.
    Chemical Name or Material Ruthenium(III) nitrosylnitrate

    H272-H314-H318-H290: May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit