Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v

Catalog No. p-7049062
Click to view available options
Quantity:
50 mL
250 mL

CAS: 34513-98-9

Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34513-98-9
MDL Number MFCD00016313

Specifications

Color Red to Brown
Linear Formula Ru(NO)(NO3)3
UN Number UN3093
Concentration Ru 1.5% w/v
Density 1.07 g/mL
Solubility Information Miscible with cold water.
Formula Weight 317.09
Odor Acidic
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Ruthenium(III) nitrosylnitrate solution
Show More Show Less
Hazard Category H290-H314-H332
Hazard Statement GHS H Statement
H272-H314
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Precautionary Statement P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
EINECSNumber 252-068-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.