Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(IV) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7058715
Click to view available options
Quantity:
5 g

Ruthenium(IV) sulfide is used to prepare sintered material, which is a semiconductor. It acts as an electrode and posses high catalytic activity for oxygen evolution from aqueous solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12166-20-0
Molecular Formula RuS2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00799846
InChI Key XNQLQKWNBPVVGB-UHFFFAOYSA-N
Synonym ruthenium iv sulfide, ruthenium sulfide, bis sulfanylidene ruthenium, dithioxoruthenium, ruthenium disulphide, disulfanylideneruthenium
PubChem CID 82958
SMILES S=[Ru]=S

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN3178
IUPAC Name bis(sulfanylidene)ruthenium
Formula Weight 165.19
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Ruthenium (IV) sulfide
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 235-318-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.