Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ruthenium Red, ≥85%, pure

Catalog No. p-7077220
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 11103-72-3 | H24Cl6N14O2Ru3 | 786.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11103-72-3
Molecular Formula H24Cl6N14O2Ru3
Molecular Weight (g/mol) 786.35
InChI Key JQJSTVUROJELSR-UHFFFAOYSA-H
Synonym 6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride
PubChem CID 16218584
IUPAC Name azane;ruthenium(2+);hexachloride;dihydrate
SMILES N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2]

Specifications

Melting Point > 500.0°C
Quantity 1 g
Assay Percent Range ≥85%
Linear Formula [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
Merck Index 15,8434
Solubility Information Solubility in water: soluble.
Formula Weight 786.35
Percent Purity ≥85%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Ruthenium Red
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.