Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ruthenium Red
GSA_VAGREENER_CHOICESDP

Catalog No. 1439100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-391-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-391-00 Supplier R&D Systems™ Supplier No. 1439/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective CaV2.1 and 2.2 blocker; also blocks TRPV channels

Ruthenium Red blocks Ca2+ uptake and release from mitochondria, and Ca2+ release from ryanodine-sensitive intracellular stores. Also blocks cell membrane-located capsaicin-activated cation channels (IC50 = 14 nM) and voltage-sensitive Ca2+ channels to inhibit neurotransmitter release.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 11103-72-3
Molecular Formula Cl6H46N14O2Ru3

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 786.35
Purity >98%
Molecular Weight (g/mol) 790.374
Chemical Name or Material Ruthenium Red
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.