Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rutin trihydrate, ≥94.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187744
Click to view available options
Quantity:
25 mg
100 mg

Rutin trihydrate is a bioflavonoid, which may play a key role in the functioning of capillaries.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 250249-75-3
Physical Form Neat
Linear Formula C27H30O16 · 3H2 O
MDL Number MFCD00149490
Synonym Quercetin-3-rutinoside trihydrate; Vitamin P trihydrate
Type Analytical Standard
Quantity 25 mg
Formula Weight 664.56
Percent Purity ≥94.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.