Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ruv B Antibody (RuvB 4G5/1), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250279MFV450
Description
Ruv B Monoclonal antibody specifically detects Ruv B in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune AssaysSpecifications
| Ruv B | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| E. coli | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
| RuvB 4G5/1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmunoassay | |
| EC 3.1.22.4, Holliday junction branch migration complex subunit RuvB | |
| Ruv B protein from E. Coli | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction