Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen RUVBL1
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227147SS
SDP
View Documents
Novus Biologicals
NBP227147SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227147
SDP
View Documents
Novus Biologicals
NBP227147
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

RUVBL1 Polyclonal specifically detects RUVBL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RUVBL1
Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human
Q9Y265
8607
A portion of amino acids 400-454 of human Rvb1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers, Wnt Signaling Pathway
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
49 kDa TATA box-binding protein-interacting protein, 54 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,49 kDa TBP-interacting protein, ECP54, INO80 complex subunit H, INO80HTIP49A, NMP 238, NMP238ECP-54, Nuclear matrix protein 238, PONTIN, Pontin 52, Pontin52, RuvB (E coli homolog)-like 1, ruvB-like 1, RuvB-like 1 (E. coli), RVB1, TATA binding protein interacting protein 49 kDa, TIH1, TIP49a, TIP49TAP54-alpha, TIP60-associated protein 54-alpha
RUVBL1
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.