Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RUVBL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582708
Description
Immunogen sequence: GKTISHVIIG LKTAKGTKQL KLDPSIFESL QKERVEAGDV IYIEANSGAV KRQGRCDTYA TEFDLEAEEY VPLPKGDV.
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
Specifications
RUVBL1 | |
Polyclonal | |
Unconjugated | |
RUVBL1 | |
2510009G06Rik; 49 kDa TATA box-binding protein-interacting protein; 49 kDa TBP-interacting protein; 54 kDa erythrocyte cytosolic protein; DNA helicase p50; ECP54; ECP-54; INO80 complex subunit H; INO80H; NMP 238; NMP238; Nuclear matrix protein 238; PONTIN; Pontin 52; Pontin52; RuvB (E coli homolog)-like 1; RuvB like AAA ATPase 1; Ruvbl1; ruvB-like 1; RuvB-like AAA ATPase 1; RuvB-like protein 1; RVB1; TAP54-alpha; TATA binding protein interacting protein 49 kDa; TIH1; Tip49; TIP49A; TIP60-a; TIP60-associated protein 54-alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8607 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y265 | |
RUVBL1 | |
Recombinant protein corresponding to Human RUVBL1. Recombinant protein control fragment (Product #RP-91880). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction