Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUVBL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589672 Supplier Invitrogen™ Supplier No. PA589672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKIEEVKSTT KTQRIASHSH VKGLGLDESG LAKQAASGLV GQENAREACG VIVELIKSKK MAGRAVLLAG PPGTGKTALA LAIAQELGSK VPFCPMVGSE VYSTEIKKTE VLMENFRRAI GLRIKETKEV YEGEVTELTP CETENPMGGY GKTISHVIIG LKTAKGTKQL KLDPSIFESL QKERVEAGDV IYIEANSGAV KRQGRCDTYA TEFD; Positive Samples: MCF-7, HeLa, 293T, Mouse testis, Mouse spleen, Rat testis; Cellular Location: Cytoplasm, Membrane, Nucleus, Nucleus matrix, centrosome, cytoskeleton, microtubule organizing center, nucleoplasm.

The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUVBL1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RUVBL1
Gene Accession No. P60122, P60123, Q9Y265
Gene Alias 2510009G06Rik; 49 kDa TATA box-binding protein-interacting protein; 49 kDa TBP-interacting protein; 54 kDa erythrocyte cytosolic protein; DNA helicase p50; ECP54; ECP-54; INO80 complex subunit H; INO80H; NMP 238; NMP238; Nuclear matrix protein 238; PONTIN; Pontin 52; Pontin52; RuvB (E coli homolog)-like 1; RuvB like AAA ATPase 1; Ruvbl1; ruvB-like 1; RuvB-like AAA ATPase 1; RuvB-like protein 1; RVB1; TAP54-alpha; TATA binding protein interacting protein 49 kDa; TIH1; Tip49; TIP49A; TIP60-a; TIP60-associated protein 54-alpha
Gene Symbols RUVBL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-214 of human RUVBL1/TIP49A/PONTIN (NP_0036981).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56505, 65137, 8607
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.