Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUVBL2 Monoclonal Antibody (A1E12)
GREENER_CHOICE

Catalog No. PIMA536098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536098 Supplier Invitrogen™ Supplier No. MA536098
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RUVBL2 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Possesses single-stranded DNA-stimulated ATPase and ATP-dependent DNA helicase (5' to 3') activity. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. RUVBL2 plays an essential role in oncogenic transformation by MYC and also modulates transcriptional activation by the LEF1/TCF1-CTNNB1 complex. May also inhibit the transcriptional activity of ATF2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUVBL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone A1E12
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Ruvbl2
Gene Accession No. Q9WTM5, Q9Y230
Gene Alias 48 kDa TATA box-binding protein-interacting protein; 48 kDa TBP-interacting protein; 51 kDa erythrocyte cytosolic protein; CGI-46; ECP51; ECP-51; erythrocyte cytosolic protein, 51-KD; INO80 complex subunit J; INO80J; liebeskummer; lik; mp47; p47; p47 protein; Repressing pontin 52; reptin; Reptin 52; reptin52 protein; RuvB (E coli homolog)-like 2; RuvB like AAA ATPase 2; RuvB like AAA ATPase 2 S homeolog; ruvbl2; RUVBL2 protein; ruvbl2.S; ruvB-like 2; RuvB-like AAA ATPase 2; RuvB-like DNA helicase reptin; RuvB-like protein 2; RVB2; TAP54-beta; TIH2; TIP48; TIP49B; TIP60-associated protein 54-beta; wu:fi25f01; XELAEV_180378471mg; xReptin; zReptin
Gene Symbols Ruvbl2
Host Species Mouse
Immunogen Recombinant protein within Human RUVB2 aa 130-321.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10856, 20174, 292907
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.